siRNA Details
Gene Symbol: HFE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000410.3 | NP_000401.1 | 4 | 911 | ||
NM_001300749.1 | NP_001287678.1 | 4 | 911 | |||
NM_139003.2 | NP_620572.1 | 3 | 593 | |||
NM_139004.2 | NP_620573.1 | 3 | 635 | |||
NM_139006.2 | NP_620575.1 | 4 | 869 | |||
NM_139007.2 | NP_620576.1 | 3 | 647 | |||
NM_139008.2 | NP_620577.1 | 3 | 605 | |||
NM_139009.2 | NP_620578.1 | 4 | 842 | |||
NM_139010.2 | NP_620579.1 | 2 | 371 | |||
XM_011514543.2 | XP_011512845.1 | 4 | 873 | |||
GenBank mRNA | AF079407.1 | - | 4 | 745 | ||
AF079408.1 | - | 3 | 523 | |||
AF079409.1 | - | 3 | 481 | |||
AF115265.1 | - | 4 | 762 | |||
AF144242.1 | - | 3 | 598 | |||
AF149804.1 | - | 3 | 477 | |||
AJ249335.1 | - | 4 | 842 | |||
AJ249336.1 | - | 3 | 647 | |||
AJ249337.1 | - | 3 | 635 | |||
AJ250635.1 | - | 2 | 371 | |||
AK293393.1 | - | 3 | 972 | |||
AK300933.1 | - | 4 | 755 | |||
AK316502.1 | - | 4 | 924 | |||
AY205604.1 | - | 3 | 512 | |||
BC074721.2 | - | 5 | 791 | 30915171 | ||
BC117201.1 | - | 4 | 786 | 40125752 | ||
BC117203.1 | - | 4 | 786 | 40125754 | ||
U60319.1 | - | 4 | 972 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
