siRNA Details
Gene Symbol: HLA-DQA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF012768.1 | - | 1 | 182 | ||
AK130598.1 | - | 2 | 333 | |||
AK130838.1 | - | 2 | 335 | |||
BC008585.1 | - | 2 | 316 | 4339446 | ||
L34091.1 | - | 1 | 213 | |||
L34092.1 | - | 1 | 213 | |||
L34093.1 | - | 1 | 213 | |||
M20506.1 | - | 1 | 181 | |||
X00370.1 | - | 2 | 292 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
