siRNA Details
Gene Symbol: RGS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276260.1 | NP_001263189.1 | 1 | 331 | ||
NM_001276261.1 | NP_001263190.1 | 10 | 1233 | |||
NM_001282923.1 | NP_001269852.1 | 17 | 1799 | |||
NM_001322214.1 | NP_001309143.1 | 10 | 1233 | |||
NM_017790.4 | NP_060260.3 | 17 | 1693 | |||
NM_130795.3 | NP_570613.2 | 10 | 1233 | |||
NM_144488.5 | NP_652759.3 | 20 | 2137 | |||
XM_006717219.2 | XP_006717282.1 | 19 | 1983 | |||
XM_006717220.1 | XP_006717283.1 | 17 | 1759 | |||
XM_006717222.1 | XP_006717285.1 | 10 | 1109 | |||
XM_011518893.1 | XP_011517195.1 | 20 | 2135 | |||
XM_011518900.1 | XP_011517202.1 | 1 | 12 | |||
XM_017015004.1 | XP_016870493.1 | 16 | 1507 | |||
XM_017015005.1 | XP_016870494.1 | 4 | 346 | |||
GenBank mRNA | AF463495.1 | - | 10 | 1233 | ||
AF490839.1 | - | 17 | 1614 | |||
AF490840.1 | - | 10 | 1083 | |||
AK000377.1 | - | 9 | 1210 | |||
AK074977.1 | - | 17 | 1663 | |||
AK096840.1 | - | 10 | 1153 | |||
AK123387.1 | - | 1 | 304 | |||
AK125059.1 | - | 17 | 1770 | |||
AK128127.1 | - | 20 | 2135 | |||
AK222888.1 | - | 9 | 1210 | |||
BC042555.1 | - | 10 | 1138 | 5175121 | ||
DA396065.1 | - | 1 | 331 | |||
FJ423754.1 | - | 62 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
