siRNA Details
Gene Symbol: RGS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322214.1 | NP_001309143.1 | 10, 11 | 1348 | ||
NM_017790.4 | NP_060260.3 | 17, 18 | 1808 | |||
XM_011518893.1 | XP_011517195.1 | 20, 21 | 2250 | |||
GenBank mRNA | AK000377.1 | - | 9, 10 | 1325 | ||
AK074977.1 | - | 17, 18 | 1778 | |||
AK222888.1 | - | 9, 10 | 1325 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
