siRNA Details
Gene Symbol: TNFRSF25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003790.2 | NP_003781.1 | 9 | 906 | ||
NM_148965.1 | NP_683866.1 | 9 | 933 | |||
NM_148966.1 | NP_683867.1 | 8 | 795 | |||
NM_148967.1 | NP_683868.1 | 8 | 771 | |||
NM_148970.1 | NP_683871.1 | 4 | 357 | |||
GenBank mRNA | AF026070.1 | - | 9 | 913 | ||
AF026071.1 | - | 8 | 1007 | |||
AK094463.1 | - | 3 | 872 | |||
AY358309.1 | - | 9 | 920 | |||
BC117189.1 | - | 9 | 886 | 40125740 | ||
BC143886.1 | - | 8 | 775 | 9052402 | ||
U72763.1 | - | 9 | 818 | |||
U74611.1 | - | 9 | 906 | |||
U75380.1 | - | 9 | 809 | |||
U78029.1 | - | 9 | 818 | |||
U83597.1 | - | 9 | 802 | |||
U94501.1 | - | 9 | 818 | |||
U94502.1 | - | 9 | 821 | |||
U94503.1 | - | 8 | 919 | |||
U94504.1 | - | 8 | 762 | |||
U94505.1 | - | 7 | 651 | |||
U94506.1 | - | 6 | 516 | |||
U94507.1 | - | 4 | 402 | |||
U94508.1 | - | 4 | 269 | |||
U94509.1 | - | 8 | 683 | |||
U94510.1 | - | 8 | 707 | |||
Y09392.1 | - | 8 | 987 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
