siRNA Details
Gene Symbol: TNFRSF25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003790.2 | NP_003781.1 | 10 | 1210 | ||
NM_148965.1 | NP_683866.1 | 10 | 1237 | |||
NM_148966.1 | NP_683867.1 | 9 | 1099 | |||
NM_148967.1 | NP_683868.1 | 9 | 1075 | |||
NM_148970.1 | NP_683871.1 | 5 | 661 | |||
GenBank mRNA | AF026070.1 | - | 10 | 1217 | ||
AF026071.1 | - | 9 | 1311 | |||
AK094463.1 | - | 4 | 1176 | |||
AY358309.1 | - | 10 | 1224 | |||
BC117189.1 | - | 10 | 1190 | 40125740 | ||
BC143886.1 | - | 9 | 1079 | 9052402 | ||
U72763.1 | - | 10 | 1122 | |||
U74611.1 | - | 10 | 1210 | |||
U75380.1 | - | 10 | 1113 | |||
U78029.1 | - | 10 | 1122 | |||
U83597.1 | - | 10 | 1106 | |||
U94501.1 | - | 10 | 1122 | |||
U94502.1 | - | 10 | 1125 | |||
U94503.1 | - | 9 | 1223 | |||
U94504.1 | - | 9 | 1066 | |||
U94505.1 | - | 8 | 955 | |||
U94506.1 | - | 7 | 820 | |||
U94507.1 | - | 5 | 706 | |||
U94508.1 | - | 5 | 573 | |||
U94509.1 | - | 9 | 987 | |||
U94510.1 | - | 9 | 1011 | |||
Y09392.1 | - | 9 | 1291 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
