siRNA Details
Gene Symbol: TNFSF13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172087.2 | NP_742084.1 | 2, 3 | 1086 | ||
GenBank mRNA | AF114012.1 | - | 2, 3 | 535 | ||
AK090698.1 | - | 2, 3 | 902 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
