siRNA Details
Gene Symbol: TNFRSF18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004195.2 | NP_004186.1 | 4 | 719 | ||
XM_017002722.1 | XP_016858211.1 | 4 | 1910 | |||
GenBank mRNA | AF125304.1 | - | 4 | 581 | ||
AY358877.1 | - | 4 | 701 | |||
BC152381.1 | - | 4 | 584 | 100013440 | ||
BC152386.1 | - | 4 | 584 | |||
BT019531.1 | - | 4 | 581 | |||
BT019532.1 | - | 4 | 581 | |||
KP067911.1 | - | 4 | 719 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
