siRNA Details
Gene Symbol: SSX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021014.3 | NP_066294.1 | 6, 7 | 604 | ||
GenBank mRNA | BC103862.1 | - | 6, 7 | 603 | 40003336 | |
S82471.1 | - | 5, 6 | 464 | |||
U90840.1 | - | 6, 7 | 517 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
