siRNA Details
Gene Symbol: DLC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164271.1 | NP_001157743.1 | 7 | 2021 | ||
NM_001316668.1 | NP_001303597.1 | 7 | 2209 | |||
NM_006094.4 | NP_006085.2 | 7 | 2279 | |||
NM_182643.2 | NP_872584.2 | 11 | 3714 | |||
XM_005273374.1 | XP_005273431.1 | 11 | 3595 | |||
XM_017012951.1 | XP_016868440.1 | 11 | 3465 | |||
XM_017012952.1 | XP_016868441.1 | 7 | 2012 | |||
GenBank mRNA | AB051510.1 | - | 11 | 3600 | ||
AF026219.1 | - | 7 | 2232 | |||
AF035119.1 | - | 7 | 2283 | |||
AK025544.1 | - | 7 | 2209 | |||
AK025733.1 | - | 2 | 455 | |||
AK289734.1 | - | 7 | 2202 | |||
AK291682.1 | - | 11 | 3555 | |||
AK299049.1 | - | 7 | 2021 | |||
BC054511.1 | - | 11 | 3469 | 5240161 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
