siRNA Details
Gene Symbol: DLC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164271.1 | NP_001157743.1 | 3, 4 | 258 | ||
NM_001316668.1 | NP_001303597.1 | 3, 4 | 446 | |||
NM_006094.4 | NP_006085.2 | 3, 4 | 516 | |||
NM_182643.2 | NP_872584.2 | 7, 8 | 1951 | |||
XM_005273374.1 | XP_005273431.1 | 7, 8 | 1832 | |||
XM_017012951.1 | XP_016868440.1 | 7, 8 | 1702 | |||
XM_017012952.1 | XP_016868441.1 | 3, 4 | 249 | |||
GenBank mRNA | AB051510.1 | - | 7, 8 | 1837 | ||
AF026219.1 | - | 3, 4 | 469 | |||
AF035119.1 | - | 3, 4 | 520 | |||
AK025544.1 | - | 3, 4 | 446 | |||
AK289734.1 | - | 3, 4 | 439 | |||
AK291682.1 | - | 7, 8 | 1792 | |||
AK299049.1 | - | 3, 4 | 258 | |||
BC054511.1 | - | 7, 8 | 1706 | 5240161 | ||
BP284911.1 | - | 3, 4 | 505 | |||
CB961755.1 | - | 3, 4 | 543 | |||
DA401187.1 | - | 3, 4 | 446 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
