siRNA Details
Gene Symbol: DLC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164271.1 | NP_001157743.1 | 2 | 121 | ||
NM_001316668.1 | NP_001303597.1 | 2 | 309 | |||
NM_006094.4 | NP_006085.2 | 2 | 379 | |||
NM_182643.2 | NP_872584.2 | 6 | 1814 | |||
XM_005273374.1 | XP_005273431.1 | 6 | 1695 | |||
XM_017012951.1 | XP_016868440.1 | 6 | 1565 | |||
XM_017012952.1 | XP_016868441.1 | 2 | 112 | |||
GenBank mRNA | AB051510.1 | - | 6 | 1700 | ||
AF026219.1 | - | 2 | 332 | |||
AF035119.1 | - | 2 | 383 | |||
AK025544.1 | - | 2 | 309 | |||
AK289734.1 | - | 2 | 302 | |||
AK291682.1 | - | 6 | 1655 | |||
AK299049.1 | - | 2 | 121 | |||
BC054511.1 | - | 6 | 1569 | 5240161 | ||
BP284911.1 | - | 2 | 368 | |||
CB961755.1 | - | 2 | 406 | |||
DA401187.1 | - | 2 | 309 | |||
DQ092382.1 | - | 2 | 330 | |||
DQ092383.1 | - | 2 | 155 | |||
DQ092384.1 | - | 73 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
