siRNA Details
Gene Symbol: WWP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270453.1 | NP_001257382.1 | 6 | 761 | ||
NM_001270454.1 | NP_001257383.1 | 9 | 1071 | |||
NM_001270455.1 | NP_001257384.1 | 9 | 1071 | |||
NM_007014.4 | NP_008945.2 | 10 | 1124 | |||
XM_011522823.2 | XP_011521125.1 | 11 | 1154 | |||
XM_011522825.1 | XP_011521127.1 | 9 | 1035 | |||
XM_011522826.2 | XP_011521128.1 | 6 | 925 | |||
XM_017022879.1 | XP_016878368.1 | 11 | 1228 | |||
XM_017022880.1 | XP_016878369.1 | 11 | 1160 | |||
XM_017022881.1 | XP_016878370.1 | 11 | 1204 | |||
GenBank mRNA | AK295070.1 | - | 2 | 116 | ||
AK295701.1 | - | 7 | 820 | |||
AK300266.1 | - | 6 | 761 | |||
AK312792.1 | - | 9 | 1051 | |||
BC000108.2 | - | 8 | 973 | |||
BC013645.1 | - | 8 | 958 | 3453212 | ||
BC064531.1 | - | 9 | 1010 | 5588092 | ||
JN712744.1 | - | 9 | 1041 | |||
U96114.1 | - | 10 | 1056 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
