siRNA Details
Gene Symbol: WWP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270453.1 | NP_001257382.1 | 7 | 973 | ||
NM_001270454.1 | NP_001257383.1 | 10 | 1283 | |||
NM_007014.4 | NP_008945.2 | 11 | 1336 | |||
XM_011522823.2 | XP_011521125.1 | 12 | 1366 | |||
XM_011522825.1 | XP_011521127.1 | 10 | 1247 | |||
XM_011522826.2 | XP_011521128.1 | 7 | 1137 | |||
XM_017022879.1 | XP_016878368.1 | 12 | 1440 | |||
XM_017022880.1 | XP_016878369.1 | 12 | 1372 | |||
XM_017022881.1 | XP_016878370.1 | 12 | 1416 | |||
GenBank mRNA | AK295070.1 | - | 3 | 328 | ||
AK295701.1 | - | 8 | 1032 | |||
AK312792.1 | - | 10 | 1263 | |||
AK316097.1 | - | 1 | 282 | |||
BC013645.1 | - | 9 | 1170 | 3453212 | ||
BC064531.1 | - | 10 | 1222 | 5588092 | ||
JN712744.1 | - | 10 | 1253 | |||
U96114.1 | - | 11 | 1268 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_025830, AK088936, AK146306, BC048184, JN712751, and others.
Gene Ontology Categories:
