siRNA Details
Gene Symbol: CAPN10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023083.3 | NP_075571.1 | 10 | 2120 | ||
NM_023085.3 | NP_075573.2 | 8 | 1655 | |||
GenBank mRNA | AB058748.1 | - | 10 | 2024 | ||
AF089088.1 | - | 10 | 2101 | |||
AF089090.1 | - | 10 | 1796 | |||
AF089091.1 | - | 8 | 1500 | |||
AF089092.1 | - | 9 | 1703 | |||
AF089093.1 | - | 11 | 2015 | |||
AF089094.1 | - | 10 | 1954 | |||
AF089096.1 | - | 2 | 363 | |||
AK027330.1 | - | 9 | 1389 | |||
AK074807.1 | - | 10 | 2071 | |||
AK128552.1 | - | 7 | 3917 | |||
BC004260.1 | - | 10 | 2069 | 3606959 | ||
BC007553.1 | - | 10 | 2069 | 2967328 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
