siRNA Details
Gene Symbol: CAPN10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023083.3 | NP_075571.1 | 7 | 1325 | ||
NM_023085.3 | NP_075573.2 | 7 | 1325 | |||
GenBank mRNA | AB058748.1 | - | 7 | 1229 | ||
AF089088.1 | - | 7 | 1306 | |||
AF089090.1 | - | 7 | 1170 | |||
AF089091.1 | - | 7 | 1170 | |||
AF089092.1 | - | 7 | 1170 | |||
AF089093.1 | - | 7 | 1170 | |||
AF089094.1 | - | 7 | 1159 | |||
AK027330.1 | - | 5 | 701 | |||
AK074807.1 | - | 7 | 1276 | |||
AK128552.1 | - | 4 | 2737 | |||
AK300279.1 | - | 7 | 1307 | |||
BC004260.1 | - | 7 | 1274 | 3606959 | ||
BC007553.1 | - | 7 | 1274 | 2967328 | ||
BQ962770.1 | - | 3 | 447 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
