siRNA Details
Gene Symbol: ATRIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271022.1 | NP_001257951.1 | 2 | 165 | ||
NM_001271023.1 | NP_001257952.1 | 2 | 505 | |||
NM_032166.3 | NP_115542.2 | 2 | 458 | |||
NM_130384.2 | NP_569055.1 | 2 | 458 | |||
GenBank mRNA | AF451323.1 | - | 2 | 354 | ||
AK022405.1 | - | 1 | 53 | |||
AK291829.1 | - | 2 | 389 | |||
AK300548.1 | - | 2 | 138 | |||
AK315075.1 | - | 2 | 422 | |||
BC014153.2 | - | 2 | 373 | 3677759 | ||
BC020563.2 | - | 2 | 381 | 3853306 | ||
BC030597.2 | - | 2 | 505 | 4826856 | ||
DC402185.1 | - | 2 | 165 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
