siRNA Details
Gene Symbol: ATRIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271022.1 | NP_001257951.1 | 5, 6 | 597 | ||
NM_001271023.1 | NP_001257952.1 | 4, 5 | 854 | |||
NM_032166.3 | NP_115542.2 | 4, 5 | 807 | |||
NM_130384.2 | NP_569055.1 | 4, 5 | 807 | |||
GenBank mRNA | AF451323.1 | - | 4, 5 | 703 | ||
AK022405.1 | - | 3, 4 | 402 | |||
AK291568.1 | - | 2, 3 | 273 | |||
AK291829.1 | - | 4, 5 | 738 | |||
AK300548.1 | - | 5, 6 | 570 | |||
AK315075.1 | - | 4, 5 | 771 | |||
AL832917.1 | - | 2, 3 | 272 | |||
BC014153.2 | - | 4, 5 | 722 | 3677759 | ||
BC020563.2 | - | 4, 5 | 730 | 3853306 | ||
BC030597.2 | - | 4, 5 | 854 | 4826856 | ||
DC402185.1 | - | 4, 5 | 514 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
