siRNA Details
Gene Symbol: WDFY3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014991.4 | NP_055806.2 | 10 | 1420 | ||
XM_005262858.4 | XP_005262915.1 | 12 | 1461 | |||
XM_011531757.2 | XP_011530059.1 | 11 | 1372 | |||
XM_011531759.2 | XP_011530061.1 | 10 | 1334 | |||
XM_011531760.2 | XP_011530062.1 | 10 | 1272 | |||
XM_011531761.2 | XP_011530063.1 | 11 | 1372 | |||
XM_011531762.2 | XP_011530064.1 | 11 | 1372 | |||
XM_011531763.2 | XP_011530065.1 | 9 | 1218 | |||
XM_011531764.2 | XP_011530066.1 | 11 | 1372 | |||
XM_011531765.2 | XP_011530067.1 | 8 | 1074 | |||
XM_011531766.2 | XP_011530068.1 | 11 | 1372 | |||
XM_011531767.2 | XP_011530069.1 | 11 | 1372 | |||
XM_017007906.1 | XP_016863395.1 | 13 | 1589 | |||
XM_017007907.1 | XP_016863396.1 | 13 | 1586 | |||
XM_017007908.1 | XP_016863397.1 | 12 | 1518 | |||
GenBank mRNA | AF538685.1 | - | 9 | 1312 | ||
AK092019.1 | - | 8 | 1074 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
