siRNA Details
Gene Symbol: WDFY3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014991.4 | NP_055806.2 | 4 | 536 | ||
XM_005262858.4 | XP_005262915.1 | 5 | 523 | |||
XM_011531757.2 | XP_011530059.1 | 4 | 434 | |||
XM_011531759.2 | XP_011530061.1 | 3 | 396 | |||
XM_011531760.2 | XP_011530062.1 | 3 | 334 | |||
XM_011531761.2 | XP_011530063.1 | 4 | 434 | |||
XM_011531762.2 | XP_011530064.1 | 4 | 434 | |||
XM_011531763.2 | XP_011530065.1 | 3 | 334 | |||
XM_011531764.2 | XP_011530066.1 | 4 | 434 | |||
XM_011531765.2 | XP_011530067.1 | 2 | 190 | |||
XM_011531766.2 | XP_011530068.1 | 4 | 434 | |||
XM_011531767.2 | XP_011530069.1 | 4 | 434 | |||
XM_017007906.1 | XP_016863395.1 | 6 | 651 | |||
XM_017007907.1 | XP_016863396.1 | 6 | 648 | |||
XM_017007908.1 | XP_016863397.1 | 5 | 580 | |||
GenBank mRNA | AF538685.1 | - | 3 | 428 | ||
AI962188.1 | - | 3 | 402 | |||
AK092019.1 | - | 2 | 190 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
