siRNA Details
Gene Symbol: VSX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014588.5 | NP_055403.2 | 5 | 1218 | ||
GenBank mRNA | AF176797.2 | - | 5 | 1208 | ||
AF251033.1 | - | 5 | 970 | |||
BC126228.1 | - | 5 | 960 | 8991944 | ||
BC136497.1 | - | 5 | 960 | 9020486 | ||
DQ854807.1 | - | 5 | 970 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
