siRNA Details
Gene Symbol: POLR1D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206559.1 | NP_001193488.1 | 3 | 514 | ||
NM_152705.2 | NP_689918.1 | 3 | 347 | |||
XM_005266414.2 | XP_005266471.1 | 4 | 1187 | |||
XM_017020622.1 | XP_016876111.1 | 2 | 214 | |||
XM_017020623.1 | XP_016876112.1 | 4 | 1107 | |||
GenBank mRNA | AF131855.1 | - | 1 | 1100 | ||
AK097973.1 | - | 4 | 434 | |||
AK307356.1 | - | 3 | 311 | |||
AK311833.1 | - | 3 | 234 | |||
BC006972.1 | - | 3 | 247 | |||
BC015319.2 | - | 3 | 256 | 3847651 | ||
BC018528.1 | - | 3 | 260 | 3865616 | ||
DB455307.1 | - | 3 | 347 | |||
DB459107.1 | - | 3 | 304 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
