siRNA Details
Gene Symbol: POLR1D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015972.3 | NP_057056.1 | 2 | 537 | ||
XM_005266412.1 | XP_005266469.1 | 3 | 1377 | |||
GenBank mRNA | AF077044.1 | - | 2 | 425 | ||
AK000623.1 | - | 2 | 423 | |||
BC000889.1 | - | 2 | 441 | 3462906 | ||
BI253733.1 | - | 2 | 349 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
