siRNA Details
Gene Symbol: CKLF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040138.2 | NP_001035228.1 | 2, 3 | 389 | ||
NM_016951.3 | NP_058647.1 | 2, 3 | 389 | |||
GenBank mRNA | AF057306.1 | - | 2, 3 | 367 | ||
AF135380.2 | - | 2, 3 | 383 | |||
AF151058.1 | - | 2, 3 | 386 | |||
AK301240.1 | - | 2, 3 | 380 | |||
AY358999.1 | - | 2, 3 | 366 | |||
BC004380.1 | - | 2, 3 | 372 | 3639550 | ||
DA804889.1 | - | 2, 3 | 370 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
