siRNA Details
Gene Symbol: ANKHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001197030.1 | NP_001183959.1 | 10 | 1869 | ||
NM_017978.2 | NP_060448.1 | 11 | 1946 | |||
NM_024668.3 | NP_078944.2 | 11 | 1979 | |||
GenBank mRNA | AF258557.1 | - | 11 | 1939 | ||
AK000295.1 | - | 3 | 252 | |||
AK022041.1 | - | 10 | 1515 | |||
AK074173.1 | - | 11 | 1943 | |||
AY070436.1 | - | 3 | 252 | |||
BC004457.1 | - | 11 | 1924 | |||
BC009420.1 | - | 11 | 1839 | |||
BC009909.2 | - | 11 | 1924 | |||
BC117677.2 | - | 11 | 1905 | |||
BC117678.2 | - | 10 | 1795 | |||
BC127127.1 | - | 11 | 1905 | |||
BC150486.2 | - | 11 | 1909 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
