siRNA Details
Gene Symbol: LYNX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023946.3 | NP_076435.1 | 4 | 428 | ||
NM_177458.1 | NP_803253.1 | 2 | 94 | |||
GenBank mRNA | AB081838.1 | - | 2 | 94 | ||
AY358417.1 | - | 2 | 110 | |||
AY587277.1 | - | 2 | 72 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
