siRNA Details
Gene Symbol: LYNX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177457.4 | NP_803252.1 | 4 | 472 | ||
NM_177476.3 | NP_803429.1 | 4 | 491 | |||
NM_177477.3 | NP_803430.1 | 4 | 605 | |||
GenBank mRNA | AF321824.1 | - | 4 | 431 | ||
BC014560.2 | - | 2 | 887 | |||
BG701653.1 | - | 4 | 491 | |||
BI603715.1 | - | 4 | 609 | |||
BM172253.1 | - | 4 | 490 | |||
BQ685632.1 | - | 4 | 420 | |||
HY110957.1 | - | 4 | 489 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
