siRNA Details
Gene Symbol: TRIM7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_203293.2 | NP_976038.1 | 7 | 1258 | ||
NM_203294.1 | NP_976039.1 | 7 | 872 | |||
NM_203295.1 | NP_976040.1 | 7 | 940 | |||
NM_203296.1 | NP_976041.1 | 7 | 1149 | |||
NM_203297.1 | NP_976042.1 | 5 | 1306 | |||
XM_017009903.1 | XP_016865392.1 | 7 | 1297 | |||
GenBank mRNA | AF396651.1 | - | 7 | 1135 | ||
AF396652.1 | - | 7 | 872 | |||
AF396653.1 | - | 7 | 940 | |||
AF396654.1 | - | 7 | 933 | |||
AF396655.1 | - | 5 | 1312 | |||
AK124617.1 | - | 2 | 2659 | |||
BC132863.1 | - | 7 | 1137 | 40146885 | ||
BC132867.1 | - | 7 | 1137 | 40146889 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
