siRNA Details
Gene Symbol: TRIM7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033342.3 | NP_203128.1 | 1 | 286 | ||
NM_203293.2 | NP_976038.1 | 1 | 286 | |||
XM_017009904.1 | XP_016865393.1 | 1 | 993 | |||
GenBank mRNA | AF220032.1 | - | 1 | 286 | ||
AF396651.1 | - | 1 | 163 | |||
BC011567.1 | - | 1 | 139 | |||
BC080553.1 | - | 1 | 116 | |||
BC132863.1 | - | 1 | 165 | 40146885 | ||
BC132867.1 | - | 1 | 165 | 40146889 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
