siRNA Details
Gene Symbol: HSFY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152584.1 | NP_689797.1 | 2 | 669 | ||
NR_003510.1 | 6 | 1107 | ||||
GenBank mRNA | AF332226.1 | - | 2 | 669 | ||
AJ566404.1 | - | 6 | 1107 |
Gene Symbol: HSFY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001877.1 | NP_001001877.1 | 2 | 669 | ||
NR_003509.1 | 6 | 1127 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
