siRNA Details
Gene Symbol: APOBEC3F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145298.5 | NP_660341.2 | 7 | 1362 | ||
XM_011529994.2 | XP_011528296.1 | 6 | 1633 | |||
GenBank mRNA | BC038808.1 | - | 7 | 1147 | 5227163 | |
BE892372.1 | - | 2 | 268 | |||
BG744406.1 | - | 2 | 258 | |||
BX367450.1 | - | 3 | 388 | |||
JF262037.1 | - | 7 | 1069 | |||
KC329530.1 | - | 7 | 1069 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
