siRNA Details
Gene Symbol: DEFB119 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271209.1 | NP_001258138.1 | 2 | 299 | ||
NM_153323.4 | NP_697018.1 | 2 | 302 | |||
GenBank mRNA | AF479699.1 | - | 2 | 133 | ||
AY122473.1 | - | 2 | 136 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
