siRNA Details
Gene Symbol: SPDYE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175064.2 | NP_778234.2 | 3, 4 | 690 | ||
XM_005249719.3 | XP_005249776.1 | 5, 6 | 1201 | |||
XM_017012015.1 | XP_016867504.1 | 5, 6 | 1139 | |||
XM_017012016.1 | XP_016867505.1 | 4, 5 | 551 | |||
XM_017012017.1 | XP_016867506.1 | 2, 3 | 786 | |||
GenBank mRNA | AF412027.1 | - | 3, 4 | 699 | ||
AL137266.1 | - | 3, 4 | 472 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
