siRNA Details
Gene Symbol: MXRA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008528.2 | NP_001008528.1 | 4 | 604 | ||
XM_005257382.2 | XP_005257439.1 | 3 | 690 | |||
XM_011524830.1 | XP_011523132.1 | 4 | 490 | |||
GenBank mRNA | AK123486.1 | - | 1 | 608 | ||
BE502793.1 | - | 4 | 455 | |||
CK300836.1 | - | 4 | 314 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
