siRNA Details
Gene Symbol: Ctsq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029636.3 | NP_083912.2 | 6 | 789 | ||
GenBank mRNA | AF456461.1 | - | 6 | 820 | ||
AK005416.1 | - | 2 | 610 | |||
AY014776.1 | - | 6 | 789 | |||
BC099415.1 | - | 6 | 796 | 30786803 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
