siRNA Details
Gene Symbol: Dnal1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028821.3 | NP_083097.2 | 6 | 379 | ||
GenBank mRNA | AK005826.1 | - | 6 | 465 | ||
BC070454.1 | - | 336 | ||||
BC125392.1 | - | 5 | 361 | 40129907 | ||
BC125394.1 | - | 5 | 357 | 40129909 | ||
BY126068.1 | - | 3 | 175 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
