siRNA Details
Gene Symbol: Fam19a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134096.2 | NP_598857.1 | 1 | 227 | ||
GenBank mRNA | AY325124.1 | - | 1 | 228 | ||
BC015306.1 | - | 1 | 228 | 4223181 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
