siRNA Details
Gene Symbol: Kctd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142731.1 | NP_001136203.1 | 3 | 2102 | ||
NM_134112.5 | NP_598873.2 | 3 | 418 | |||
GenBank mRNA | AK016623.1 | - | 2 | 463 | ||
AK054557.1 | - | 3 | 409 | |||
AY899808.1 | - | 3 | 418 | |||
BC022615.1 | - | 1 | 86 | |||
BC087623.1 | - | 3 | 317 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
