siRNA Details
Gene Symbol: Snrpd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026943.1 | NP_081219.1 | 3 | 401 | ||
GenBank mRNA | AK007389.1 | - | 3 | 380 | ||
AK045968.1 | - | 3 | 376 | |||
AK088105.1 | - | 3 | 380 | |||
BC043014.1 | - | 3 | 401 | 5683364 | ||
BC051208.1 | - | 2 | 380 | 1510565 | ||
BC116858.1 | - | 3 | 384 | 40126177 | ||
BC116860.1 | - | 3 | 384 | 40126179 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
