siRNA Details
Gene Symbol: Ibtk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081282.2 | NP_001074751.1 | 12 | 2718 | ||
GenBank mRNA | AK030895.1 | - | 12 | 2681 | ||
AK031796.1 | - | 1 | 519 | |||
AK050176.1 | - | 12 | 2711 | |||
AK129353.1 | - | 9 | 1596 | |||
BC050255.1 | - | 1 | 23 | |||
BC060722.1 | - | 11 | 2512 | |||
BC137798.1 | - | 12 | 2607 | 8860817 |
