siRNA Details
Gene Symbol: Ibtk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081282.2 | NP_001074751.1 | 19 | 3342 | ||
GenBank mRNA | AK031796.1 | - | 8 | 1143 | ||
AK050176.1 | - | 19 | 3335 | |||
AK129353.1 | - | 16 | 2220 | |||
BC050255.1 | - | 7 | 2776 | |||
BC060722.1 | - | 18 | 3136 | |||
BC137798.1 | - | 19 | 3231 | 8860817 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
