siRNA Details
Gene Symbol: Rcc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173867.5 | NP_776292.1 | 4 | 577 | ||
GenBank mRNA | AK076040.1 | - | 4 | 508 | ||
AK076046.1 | - | 808 | ||||
AK129370.1 | - | 3 | 281 | |||
BC020385.1 | - | 2 | 110 | |||
BC060624.1 | - | 4 | 531 | |||
BC086666.1 | - | 4 | 500 | 30634817 | ||
CA751510.1 | - | 4 | 576 |

More Information
Other Species Targeted

Gene Ontology Categories:
