siRNA Details
Gene Symbol: Apol7c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175391.4 | NP_780600.1 | 5 | 449 | ||
GenBank mRNA | AK075855.1 | - | 5 | 451 | ||
BC139148.1 | - | 5 | 408 | 8862170 | ||
BC139149.1 | - | 5 | 408 | 8862171 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
