siRNA Details
Gene Symbol: Hlcs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139145.4 | NP_631884.1 | 9 | 1977 | ||
GenBank mRNA | AB066227.1 | - | 8 | 1837 | ||
AK052424.1 | - | 10 | 2273 | |||
AK158212.1 | - | 10 | 2094 | |||
AK168925.1 | - | 6 | 872 | |||
BC050090.1 | - | 9 | 1978 | 30012868 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
