siRNA Details
Gene Symbol: Prss3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011645.2 | NP_035775.1 | 5 | 692 | ||
GenBank mRNA | AK131862.1 | - | 5 | 693 | ||
BC098498.1 | - | 5 | 675 | |||
BC141304.1 | - | 5 | 757 | 9055916 |
Gene Symbol: Gm10334 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001103153.1 | NP_001096623.1 | 5 | 708 |
Gene Symbol: Prss1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053243.2 | NP_444473.1 | 5 | 689 | ||
GenBank mRNA | BC094921.1 | - | 5 | 678 | 6433981 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
