siRNA Details
Gene Symbol: Prss1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053243.2 | NP_444473.1 | 2 | 108 | ||
GenBank mRNA | BC094921.1 | - | 2 | 97 | 6433981 |
Gene Symbol: Gm5409 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003664.2 | NP_001003664.1 | 2 | 98 |
Gene Symbol: Gm5771 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038997.2 | NP_001034086.1 | 2 | 125 | ||
GenBank mRNA | BC132026.1 | - | 2 | 125 | 40130303 | |
BC132030.1 | - | 2 | 125 | 40130307 |
Gene Symbol: Try10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038996.2 | NP_001034085.1 | 2 | 182 | ||
GenBank mRNA | AK133516.1 | - | 2 | 109 | ||
BC138698.1 | - | 2 | 182 | 8861720 | ||
BC146017.1 | - | 2 | 182 | 40131320 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
