siRNA Details
Gene Symbol: Il22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016971.2 | NP_058667.1 | 1 | 188 | ||
GenBank mRNA | AJ249491.1 | - | 1 | 214 | ||
BC104348.1 | - | 1 | 188 | 40049545 | ||
BC104349.1 | - | 1 | 188 | 40049546 | ||
BC116235.2 | - | 1 | 184 | 40091712 |
Gene Symbol: Iltifb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054079.2 | NP_473420.2 | 1 | 210 | ||
GenBank mRNA | AJ249492.1 | - | 1 | 210 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
