siRNA Details
Gene Symbol: Ank3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009670.4 | NP_033800.2 | 18 | 3147 | ||
NM_146005.3 | NP_666117.2 | 41 | 6206 | |||
NM_170687.3 | NP_733788.2 | 18 | 3735 | |||
NM_170688.2 | NP_733789.1 | 41 | 5618 | |||
NM_170689.2 | NP_733790.2 | 40 | 6152 | |||
NM_170690.2 | NP_733791.2 | 40 | 6143 | |||
NM_170728.2 | NP_733924.2 | 39 | 5501 | |||
NM_170729.2 | NP_733925.2 | 39 | 6089 | |||
NM_170730.2 | NP_733926.2 | 16 | 2837 | |||
GenBank mRNA | AF212924.1 | - | 16 | 2337 | ||
AK033389.1 | - | 3 | 1033 | |||
AK035681.1 | - | 18 | 3463 | |||
BC021657.1 | - | 39 | 5371 | 4188590 | ||
BC026152.1 | - | 18 | 2749 | |||
BC062122.1 | - | 7 | 3455 | |||
L40631.1 | - | 18 | 3738 | |||
L40632.1 | - | 41 | 5748 | |||
U89274.1 | - | 20 | 2764 | |||
U89275.1 | - | 20 | 3352 |

More Information
Other Species Targeted

Gene Ontology Categories:
