siRNA Details
Gene Symbol: Ap3s1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF084575.1 | - | 6 | 568 | ||
AK137489.1 | - | 6 | 589 | |||
AK152100.1 | - | 6 | 600 | |||
AK184535.1 | - | 4 | 248 | |||
AK194047.1 | - | 4 | 350 | |||
BC012656.1 | - | 6 | 579 | 4187607 | ||
BC108327.1 | - | 6 | 587 | 30713247 | ||
BC131985.1 | - | 6 | 569 | 40130262 | ||
BC145234.1 | - | 7 | 610 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001106933, XM_006254693, XM_006254694, XM_008772548, XM_006254695, and others.
