siRNA Details
Gene Symbol: Ap3s1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF084575.1 | - | 5 | 502 | ||
AK002565.1 | - | 5 | 528 | |||
AK137489.1 | - | 5 | 523 | |||
AK152100.1 | - | 5 | 534 | |||
AK184535.1 | - | 3 | 182 | |||
BC012656.1 | - | 5 | 513 | 4187607 | ||
BC108327.1 | - | 5 | 521 | 30713247 | ||
BC131985.1 | - | 5 | 503 | 40130262 | ||
BC145234.1 | - | 5 | 503 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
